Essential Oil on SLB®-5ms
GC Analysis of <I>Cinnamoµm camphora</I> Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 C, 3 C/min to 250 C (5 min)
carrier gas helium, 30 cm/sec, constant
sample cinnamoµm camphora essential oil diluted 1:10 in n-hexane
injection 1 µL, 50:1 split
inj. temp. 280 C
detector FlD, 280 C
Description
Analysis Note Cinnamoµm Camphor, also known as the camphor tree, is grown in several parts of the world including Japan, Taiwan, and Australia. Camphor, which is used in medicinal and cosmetic preparations, is derived from the leaves and wood of this tree. This application illustrates the GC analysis of the volatiles of cinnamoµm camphor on the SLB-5ms.
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-Caryophyllene
  • (E,E)-α-Farnesene
  • Allocimene
  • Cadin-4-en-10-ol
  • Camphene
  • Germacrene D
  • Limonene
  • Linalool
  • Menthol
  • Myrcene
  • p-Cymene
  • Spathulenol
  • Terpinen-4-ol
  • trans-Carveol
  • trans-Pinocarveol
  • Viridiflorol
  • α-Pinene
  • α-Terpineol
  • α-Terpinyl acetate
  • α-Thujene
  • β-Bourbonene
  • γ-Terpinene