GC Analysis of Ibuprofen Enantiomers (Underivatized) on β-DEX™ 120
GC Analysis of Ibuprofen Enantiomers (Underivatized) on β-DEX™ 120
Conditions
column Supelco beta-DEX 120, 30 m x 0.25 mm I.D., 0.25 µm (24304)
carrier gas helium, 60 m/sec., constant flow
gradient 50 C (5 min), 5 C/min to 200 C (10 min)
sample 2 mg/mL in methanolpeak 1: S(+)-ibuprofenpeak 2: R(-)-ibuprofen
injection 1 µL, 100:1 split
liner 4 mm I.D., FocusLiner(TM)
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Pharmaceuticals
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Ibuprofen