GC Analysis of Impurities in MTBE on Petrocol® DH Octyl
GC Analysis of Impurities in MTBE on Petrocol® DH Octyl
Conditions
column Petrocol DH Octyl, 100 m x 0.25 mm I.D., 0.50 µm (24282)
oven 35 °C (15 min), 1 °C/min to 200 °C (15 min)
carrier gas helium, 24 cm/sec @ 35 °C
injection 0.1 µL, 215:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2-Methylbutene
  • ETBE
  • Isopentane
  • Methanol
  • MTBE
  • TAME
  • tert-Butanol