GC Analysis of Industrial Solvents on SLB®-IL60i
GC Analysis of Industrial Solvents on SLB®-IL60i
Conditions
column SLB-IL60i, 30 m x 0.25 mm I.D., 0.20 µm (29832-U)
oven 40 °C (4 min), 8 °C/min to 200 °C (5 min)
carrier gas helium, 30 cm/sec
sample industrial solvents, each at 0.2% (v/v) in pentane
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note The i-series of ionic liquid capillary GC columns are characterized by their improved inertness for polar analytes. The analysis of a 56-component industrial solvent mix demonstrated the ability of SLB IL60i to produce great peak shapes for analytes with various functionality.
Featured Industry Chemicals and Industrial Polymers
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC