GC Analysis of Isooctyl Alcohols on Nukol™
GC Analysis of Isooctyl Alcohols on Nukol™
Conditions
column Nukol, 30 m x 0.53 mm I.D., 0.50 µm (25327)
oven 90 °C (5 min), 4 °C/min to 160 °C
carrier gas helium, 20 cm/sec
sample 2% isooctyl alcohols in methanol
injection 2 µL
inj. temp. 200 °C
detector FID, 200 °C
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information Nukol is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2-Ethylhexyl alcohol
  • Methanol
  • Oxo-isooctyl Alcohols