GC Analysis of JET-A on Equity®-1, Fast GC Analysis
GC Analysis of JET-A on Equity®-1, Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 80 °C, 50 °C/min to 325 °C
carrier gas hydrogen, 45 cm/sec constant
sample JET-A, neat
injection 0.1 µL, 300:1 split
liner 2 mm I.D., straight
inj. temp. 250 °C
detector FID, 350 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes