GC Analysis of Kennewick Peppermint Essential Oil on Equity®-1
GC Analysis of Kennewick Peppermint Essential Oil on Equity®-1
Conditions
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 75 C (8 min), then 4 C/min to 200 C (15 min)
carrier gas helium, 25cm/ sec set at 110C
sample Willamette Peppermint Oil, (A.M. Todd Sample), neat
injection 0.1 µL, 100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes