GC Analysis of L-Carvone Enantiomer in Scotch Spearmint Essential Oil on Astec® CHIRALDEX® G-TA
GC Analysis of L-Carvone Enantiomer in Scotch Spearmint Essential Oil on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 40 °C (1 min), 2 °C/min to 170 °C (15 min)
carrier gas helium, 25 cm/sec, set at 110 °C
sample Scotch Spearmint Oil, neat
injection 0.1 µL100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Ketones, Essential Oils, Enantiomers
Featured Industry Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes