GC Analysis of Lemon Essential Oil Contaminated with 5%Sweet Orange Essential Oil (Showing Camphene/Δ-3-Carene Ratio) on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Lemon Essential Oil Contaminated with 5%Sweet Orange Essential Oil (Showing Camphene/Δ-3-Carene Ratio) on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C, 30 °C/min to 85 °C, 80 °C/min to 320 °C
carrier gas hydrogen, 70 cm/sec constant
sample lemon essential oil + 5% sweet orange essential oil in hexane; the delta-3-carene/camphene value is 0.275, which exceeds the maxiµm value of 0.140 to be considered pure lemon essential oil
injection 0.4 µL in the split mode (300:1)
inj. temp. 320 °C
detector FID, 320 °C (sampling rate 12 ms)
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes