GC Analysis of Lemon Essential Oil on SLB®-IL59, 3 °C/min Oven Ramp
GC Analysis of Lemon Essential Oil on SLB®-IL59, 3 °C/min Oven Ramp
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.2 µm (28891-U)
oven 50 C, 3 C/min to 300 C
carrier gas helium, 30 cm/sec, constant
detector FID
Description
Analysis Note The volatile fraction of essential oils represents a quite complex matrix, being composed of a large variety of compounds, mainly terpenes, sesquiterpenes, alcohols and other oxygenated compounds, that cannot be all separated in a single GC analysis. The ionic liquid GC column, SLB®-IL59 showed a good resolving power toward all the classes of compounds present in the essential oils. It also provided good peak shape for both terpenes and oxygenated compounds thus proving it is a valuable polar GC column for the analyis of these types of samples. More than 50 compounds have been identified on this column by using mass spectra databases and by co-injection of standards.
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-Caryophyllene
  • (E)-β-Farnesene
  • (E)-β-Ocimene
  • (Z)-α-Bisabolene
  • (Z)-β-Ocimene
  • Camphene
  • Eucalyptol
  • Geranial
  • Geranyl acetate
  • Limonene
  • Linalool
  • Myrcene
  • Neral
  • Neryl acetate
  • p-Cymene
  • Sabinene
  • Terpinen-4-ol
  • Terpinolene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • α-trans-Bergamotene
  • β-Bisabolene
  • β-Phellandrene
  • β-Pinene
  • γ-Terpinene