Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Lemon Essential Oil on SLB®-IL59
GC Analysis of Lemon Essential Oil on SLB®-IL59
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
oven 50 C, 5 C/min to 300 C
carrier gas helium, 40 cm/sec
sample lemon essential oil diluted 1:10 in hexane
injection 1 muL100:1 split
inj. temp. 250 C
detector FID, 305 C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes