Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Lemon Essential Oil (Showing Camphene/Δ-3-Carene Ratio) on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Lemon Essential Oil (Showing Camphene/Δ-3-Carene Ratio) on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 C, 30 C/min to 85 C, 80 C/min to 320 C
carrier gas hydrogen, 70 cm/sec constant
sample lemon essential oil in hexane; the value for delta-3-carene/camphene must be <0.140 to be considered pure lemon essential oil
injection 0.4 µl in the split mode (300:1)
inj. temp. 320 C
detector FID, 320 C (sampling rate 12 ms)
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes