GC Analysis of Methane, Acetylene, and Ethylene on Carboxen®-1006 PLOT and a Packed Column (Carboxen 1004)
GC Analysis of Methane, Acetylene, and Ethylene on Carboxen®-1006 PLOT and a Packed Column (Carboxen 1004)
Conditions
column 2m x 1/16in. stainless steel packed with Carboxen® 1004 (12846)
column Carboxen-1006 PLOT, 30 m x 0.53 mm I.D. (25461)
oven 165 °C
carrier gas helium, 22 cm/sec
sample ethylene
injection 10 µLdirect
inj. temp. 230 °C
detector FID, 230C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Light Hydrocarbons, Hydrocarbons, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • Acetylene
  • Ethylene
  • Methane