Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Methylcyclohexane Methanol (MCHM) and Propylene Glycol Phenyl Ether (PPH) in Water on VOCOL® after SPME using 100 μm PDMS Fiber
GC Analysis of Methylcyclohexane Methanol (MCHM) and Propylene Glycol Phenyl Ether (PPH) in Water on VOCOL® after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix MCHM and PPH at 1 µg/L in 6 mL water, 25% sodium chloride
desorption process 2 min, 270 C (splitter open at 0.75 min)
extraction 5 min, headspace at 55 C, with agitation
SPME fiber SPME Assembly, 100 µm PDMS FS, 24 ga, Auto (Red) (57301)
column Vocol, 20 m x 0.18 mm I.D. x 1.0 µm (28463-U)
oven 50 C (1 min), 15 C/min to 220 C (5 min)
carrier gas helium, 1 ml/min constant
injection SPME
liner 0.75 mm I.D., SPME
inj. temp. 270 C
detector MSD, selected ion mode
MSD interface 220 C
scan range selected ion mode; m/z=110, 97 (MCHM) and m/z=94, 152 (PPH)
Description
Analysis Note In this application, an SPME method was developed for the analysis of two compounds involved in the chemical spill into the Elk River in West Virginia, which took place in January 2014. The method could easily detect both MCHM and PPH at a level of 1 μg/L from spiked surface water.
Categories Analytical Chromatography, SPME Applications, GC Applications, Alcohols, Environmental, Ethers, Solvents, Aromatics
Featured Industry Environmental
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPME, application for GC
Analyte
  • 1-Phenoxy-2-propanol