GC Analysis of Mineral Spirits on TCEP, Fast GC Analysis
GC Analysis of Mineral Spirits on TCEP, Fast GC Analysis
Conditions
column TCEP, 15 m x 0.10 mm I.D., 0.18 µm (28348-U)
oven 100 C
carrier gas hydrogen, 43 cm/sec
sample Low odor mineral spirits (from a local hardware store)
injection 0.04 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 220 C
detector FID, 220 C
Description
Categories Analytical Chromatography, GC Applications, Food & Beverage Analysis, Beverage Testing, Aromatics, Beer, Wine, and Spirits, Hydrocarbons, Spirits
Featured Industry Chemicals and Industrial Polymers
Other Notes Complementary products:
Syringes