GC Analysis of Native Spearmint Essential Oil on SLB®-5ms
GC Analysis of Native Spearmint Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 75 °C (8 min), then 4 °C/min to 200 °C (15 min)
carrier gas helium, 25cm/ sec set at 110C
sample Native Spearmint Oil, (A.M. Todd Sample), neat
injection 0.1 µL, 100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes