GC Analysis of Noble Gases on Carboxen®-1010 PLOT
GC Analysis of Noble Gases on Carboxen®-1010 PLOT
Conditions
column Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven 35 °C (6 min), 24 °C/min to 225 °C
carrier gas helium, 3 mL/min. @ 35 °C
detector TCD
Description
Categories Analytical Chromatography, GC Applications, Gases, Volatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes