GC Analysis of Omega 6 and Other FAMEs on Omegawax®
GC Analysis of Omega 6 and Other FAMEs on Omegawax®
Conditions
column Omegawax 320, 30 m x 0.32 mm I.D., 0.25 µm (24152)
oven 200 °C
carrier gas helium, 25 cm/sec @ 200 °C
sample 50 mg/mL animal source FAMEs in hexane
injection 1 µL, split 100:1
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • C14:0
  • C16:0
  • C18:2n6
  • C18:3n6
  • C20:2n6
  • C20:3n6
  • C20:4n6
  • C20:5n3
  • C22:4n6
  • C22:5n3
  • C22:5n6
  • C22:6n3
  • C24:0
  • C24:1
  • Methyl arachidonate
  • Methyl linoleate
  • Methyl myristate
  • Methyl palmitate
  • Methyl γ-linolenate