GC Analysis of Oxygen, Nitrogen, and Carbon Dioxide on Carboxen®-1010 PLOT
GC Analysis of Oxygen, Nitrogen, and Carbon Dioxide on Carboxen®-1010 PLOT
Conditions
column Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven 35 °C (7.0 min), 24 °C/min to 225 °C
carrier gas helium, 3.0 mL/min
sample Oxygen at 20% and carbon dioxide at 1.0% in nitrogen
injection 10 µL, direct using a sample valve, 150 °C
inj. temp. 200 °C
detector TCD, 230 °C, Methanizer, 375 °C, and FID, 230 °C in series
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes