GC Analysis of Patchouli Essential Oil on SLB®-5ms
GC Analysis of Patchouli Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C (0.5 min), then 5 °C/min to 300 °C (10 min)
carrier gas helium, 25cm/ sec set at 110C
sample Patchouli Oil, neat (W283800)
injection 0.1 µL100:1 split
liner 4 mm I.D. cup injector
inj. temp. 280 °C
detector FID, 310 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes