GC Analysis of Patchouli Essential Oil on SLB®-IL60
GC Analysis of Patchouli Essential Oil on SLB®-IL60
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.25 µm (29505-U)
oven 50 °C (0 min), then 5 °C/min to 250 °C (10 min)
carrier gas helium, 25cm/ sec set at 110C
sample Patchouli Oil, neat (W283800)
injection 0.1 µL100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes