Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of PCBs in Whole Cow Milk (Spiked) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
GC Analysis of PCBs in Whole Cow Milk (Spiked) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix 10 g whole cow's milk
extraction tube Supel(TM) QuE Non-buffered tube (4 g magnesium sulfate, 1 g sodium chloride) (55294-U)
clean-up tube Supel(TM) QuE Z-Sep 150 mg, magnesium sulfate (150 mg, part no. 63135) (55299-U)
column SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 75 C (1 min), 12 C/min to 340 C (10 min)
carrier gas hydrogen, 1.2 ml/min, constant
injection 1 muLsplitless (0.75 min)
liner 4 mm I.D. FocusLiner(TM) with taper
inj. temp. 250 C
detector mu-ECD, 340 C
Description
Analysis Note This application demonstrates the use of dispersive SPE or "QuEChERS" for the extraction and cleanup of whole cow′s milk for GC-ECD analysis of polychlorinated biphenyl (PCBs). Z-Sep sorbent was used in the cleanup step and found to reduce background and yield recoveries >70% for all PCB congeners evaluated except no. 209, which was recovered at 68%.
Extraction solvent: acetone:hexane (1:1), 20 mL
Clean-up volume: 1 mL
Categories Analytical Chromatography, Food & Beverage Analysis, SPE Applications, GC Applications, Dioxins, PCBs, PBDEs, Beverage Testing, Dioxins/PCBs/PBDEs, Milk
Featured Industry Food and Beverages
General description Reference: Stenerson, K.; Ye, M.; Halpenny, M., New Analytical Tools for the Determination of Persistent Organic Pollutants (POPs) in Fatty Food and Beverage Matrices Using QuEChERS Extraction/Cleanup and Gas Chromatography (GC) Analysis. Pittcon 2013. Sigma-Aldrich/Supelco publication T413034
Legal Information Supel is a trademark of Sigma-Aldrich Co. LLC, FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE