GC Analysis of Peppermint Essential Oil on SLB®-5ms
GC Analysis of Peppermint Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C, 3 °C/min to 300 °C
carrier gas helium, 30 cm/sec
sample Peppermint oil purchased from Misitano&Stracuzzi S.p.a. (Messina, Italy) diluted 1:10 in n-hexane
inj. temp. 220 °C
detector FID, 300 °C
detector 1 µL, 100:1 split
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes