GC Analysis of Peppermint Essential Oil on SLB®-IL59
GC Analysis of Peppermint Essential Oil on SLB®-IL59
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
oven 50 °C, 3 °C/min to 300 °C
carrier gas helium, 30 cm/sec
sample Peppermint oil purchased from Misitano&Stracuzzi S.p.a. (Messina, Italy) diluted 1:10 in n-hexane
injection 1 µL, 100:1 split
inj. temp. 220 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-Caryophyllene
  • (E)-β-Ocimene
  • (Z)-Jasmone
  • (Z)-β-Ocimene
  • 1,5-di-epi-β-Bourbonene
  • 2,5-Diethyltetrahydrofuran
  • Bicyclogermacrene
  • Camphene
  • Carvone
  • Caryophyllene oxide
  • cis-Sabinene hydrate
  • Eucalyptol
  • Germacrene D
  • iso-Menthol
  • iso-Menthone
  • iso-Menthyl acetate
  • Limonene
  • Linalool
  • Menthofuran
  • Menthol
  • Menthone
  • Menthyl acetate
  • Myrcene
  • Neomenthol
  • Neomenthyl acetate
  • Nonan-3-ol
  • Octan-3-ol
  • Octan-3-one
  • p-Cymene
  • Piperitone
  • Pseudolimonene
  • Pulegone
  • Sabinene
  • Terpinolene
  • Thymol
  • trans-Sabinene hydrate
  • trans-β-Bergamotene
  • Viridiflorol
  • α-Pinene
  • α-Terpinene
  • α-Thujene
  • β-Bourbonene
  • β-Phellandrene
  • β-Pinene
  • γ-Terpinene