GC Analysis of Permanent Gases and C2 Hydrocarbons on a Packed Column (60/80 Carboxen® 1000)
GC Analysis of Permanent Gases and C2 Hydrocarbons on a Packed Column (60/80 °Carboxen® 1000)
Conditions
column 15 ft. x 1/8 in. x 2.1 mm I.D., stainless steel
packing 60/80 °Carboxen 1000
oven 35 °C (5 min.), 20 °C/min. to 225 °C
carrier gas helium, 30 mL/min
sample Scott Gas mix (23437) with added oxygen
injection 0.6 mL, on-column
detector TCD
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Light Hydrocarbons, Gases, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acetylene
  • Carbon dioxide
  • Carbon monoxide
  • Ethane
  • Ethylene
  • Methane
  • Nitrogen
  • Oxygen