GC Analysis of Pesticides in Spaghetti Sauce on SLB®-5ms after QuEChERS Extraction then Cleanup using Supel QuE PSA/C18
GC Analysis of Pesticides in Spaghetti Sauce on SLB®-5ms after QuEChERS Extraction then Cleanup using Supel QuE PSA/C18
Conditions
sample preparation Concentrate 2 mL aliquot of cleaned extract to a final volume of 0.2 mL.
sample/matrix Sample pretreatment: 10 g sample spiked at 10 ng/g; Sample:spaghetti sauce, marinara
extraction tube Supel QuE Acetate (AC) tube (55234-U)
extraction process Add 10 mL 1% acetic acid in acetonitrile, shake for 10 min at 2500 rpm. Centrifuge for 5 min at 5000 rpm.
clean-up process Take 8 mL of extract and add to cleanup tube. Shake for 1 min and centrifuge at 3500 rpm for 3 min.
clean-up tube Supel QuE PSA/C18, 12 mL (55283-U)
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C (3 min), 10 °C/min to 325 °C ( 5 min)
carrier gas helium, 1 mL/min constant flow
injection 1 µL, splitless
liner 4 mm I.D. FocusLiner™ with taper
inj. temp. 250 °C
detector 325 °C
scan range SIM
Description
Application In this application, QuEChERS extraction was applied to the analysis of pesticide residues in tomato-based spaghetti sauce. Sauce spiked at 10 ng/g with a variety of pesticides was extracted following AOAC Official Method 2007.01. The resulting extract was then subjected to cleanup using PSA/C18, concentrated, and analyzed by GC/MS-SIM. Recoveries for spiked replicates were >70% for all pesticides analyzed with %RSD values <15%.
Categories Analytical Chromatography, SPE Applications, GC Applications, Pesticides
Featured Industry Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC, application for SPE