Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Petitgrain Essential Oil on Equity®-1
GC Analysis of Petitgrain Essential Oil on Equity®-1
Conditions
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 75 C (8 min), then 4 C/min to 200 C (15 min)
carrier gas helium, 25cm/ sec set at 110C
sample Petitgrain Oil, Paraguay, neat (W523607)
injection 0.1 µL, 100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes