GC Analysis of Petitgrain Essential Oil on Equity®-1, 50 °C Initial Oven Temp.
GC Analysis of Petitgrain Essential Oil on Equity®-1, 50 °C Initial Oven Temp.
Conditions
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 50 °C (0 min), then 3 °C/min to 250 °C (10 min)
carrier gas helium, 25cm/ sec set at 110C
sample Petitgrain Oil, Paraguay, neat (W523607)
injection 0.1 µL, 100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes