Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Conditions
sample/matrix Phenols at 25 ppb in 0.1M phosphate buffer, pH 2, with 25% NaCl, 9 mL in 10 mL screw cap vial
desorption process (30 min. direct immersion with agitation using CombiPal autosampler)
extraction 30 min direct immersion with agitation using CombiPal autosampler
SPME fiber 60 µm Carbowax (PEG) (57354-U)
column SLB-5ms, 30 m x 0.25 mm I.D., 0.5µm film
oven 50C(2.0 min) to 160C @ 10C/min to 280C @ 20C/min
carrier gas Helium, 1mL/min constant flow, 8.7 psi
inj. temp. Splitless/split with 0.75mm ID liner and Merlin Microseal
detector ITMS m/z=45-275 @ 0.7 musec per scan, transfer line 310C
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Phenols
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information Carbowax is a registered trademark of Union Carbide, SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Analyte
  • 2,3,4,6-Tetrachlorophenol
  • 2,4,5-Trichlorophenol
  • 2,4,6-Trichlorophenol
  • 2,4-Dichlorophenol
  • 2,4-Dimethylphenol
  • 2,4-Dinitrophenol
  • 2,6-Dichlorophenol
  • 2-Chlorophenol
  • 2-Methyl-4,6-dinitrophenol
  • 2-Methylphenol
  • 2-Nitrophenol
  • 2-sec-Butyl-4,6-dinitrophenol
  • 3-Methylphenol
  • 4-Chloro-3-methylphenol
  • 4-Methylphenol
  • 4-Nitrophenol
  • Naphthalene-d8
  • Pentachlorophenol
  • Phenol