Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Phthalate Esters in Extra Virgin Olive Oil (Spiked) on SLB®-5ms after SPME using 100 μm PDMS Fiber
GC Analysis of Phthalate Esters in Extra Virgin Olive Oil (Spiked) on SLB®-5ms after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 0.5 g extra virgin olive weighed into a 10 mL glass centrifuge tube, spiked with phthalate esters at 2 mg/kg, 3 mL acetonitrile added (vortexed 30 sec, centrifuged 10 min at 3000 rpm, 1.5 mL acetonitrile from the top transferred to a 2 mL vial)
desorption process 280 C for 5 min, splitless
extraction direct-immersion, 20 min, constant stirring at 500 rpm
SPME fiber SPME Assembly, 100µm PDMS FS, 24 GA Manual (57300-U)
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 90 C (5 min), 30 C/min to 310 C, 50 C/min to 350 C (3 min)
carrier gas helium, 50 cm/sec
inj. temp. 280 C
detector MS (triple quadrupole), ion source: 220 C; ionization mode: EI (70 eV); acquisition mode: MRM
MSD interface 280 C
Description
Analysis Note Data provided by Giorgia Purcaro (1), Laura Barp (2), and Luigi Mondello (1,2) of (1) Chromaleont s.r.l., c/o “Scienze del Farmaco e Prodotti per la Salute” Department, University of Messina, viale Annunziata, 98168 Messina, Italy or (2) “Scienze del Farmaco e Prodotti per la Salute” Department, University of Messina, viale Annunziata, 98168 Messina, Italy.
This method uses SPME with minimal sample manipulation and permits robust, fast, and accurate GC-triple quadrupole MS (QqQ) determination of phthalate esters in olive oil after their separation on an SLB-5ms column.
Categories Analytical Chromatography, GC Applications, Phthalate Esters
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPME, application for GC