GC Analysis of α-Pinene Enantiomers on Astec® CHIRALDEX® G-DP
GC Analysis of α-Pinene Enantiomers on Astec® CHIRALDEX® G-DP
Conditions
column Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 µm (78033AST)
oven 50 °C
carrier gas helium, 30 psi
sample 4 mg/mL in methylene chloridepeak 1: d-alpha-pinene (R(+)-alpha-pinene)peak 2: l-alpha-pinene (S(-)-alpha-pinene)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Terpenes
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes