Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX® G-DP
GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX® G-DP
Conditions
column Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 µm (78033AST)
oven 70 C
carrier gas helium, 30 psi
sample 5 mg/mL in ethanolpeak 1: l-beta-pinenepeak 2: d-beta-pinene
injection 0.2 µL, 80:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Analysis Note Elution order reversal on CHIRALDEX B-DM and CHIRALDEX B-PM.
Categories Analytical Chromatography, GC Applications, Enantiomers, Terpenes
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes