GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX® B-PM
GC Analysis of β-Pinene Enantiomers on Astec® CHIRALDEX® B-PM
Conditions
column Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 µm (76023AST)
oven 50 °C
carrier gas helium, 30 psi
sample 5 mg/mL in ethanolpeak 1: d-beta-pinenepeak 2: l-beta-pinene
injection 0.2 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Terpenes
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes