Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of α-Pinene Enantiomers on Astec® CHIRALDEX® B-DM
GC Analysis of α-Pinene Enantiomers on Astec® CHIRALDEX® B-DM
Conditions
column Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 µm (77023AST)
oven 50 C
carrier gas helium, 30 psi
sample 4 mg/mL in methylene chloridepeak 1: l-alpha-pinene (S(-)-alpha-pinene)peak 2: d-alpha-pinene (R(+)-alpha-pinene)
injection 1 µL, 80:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Analysis Note Elution order reversal (d,l) on CHIRALDEX G-DP.
Categories Analytical Chromatography, GC Applications, Terpenes, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes