GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-18
GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-18
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix water spiked with PAHs
SPE tube/cartridge Supelclean ENVI-18, 500 mg/6 mL (57064)
condition 2 x 6 mL toluene:methanol (10:1); 6 mL methanol; 6 mL DI water
sample addition 250 mL water sample at 10 mL/min
drying 10 min under vacuum
elution 2 x 1 mL toluene:methanol (10:1)
column 5% phenyl polysiloxane, 30m x 0.25 mm I.D., 0.25 µm film (Supelco equivalent, SLB-5ms, 28471-U)
oven 70 °C (2 min) to 280 °C at 8 °C/min
carrier gas helium
injection 1 µL
detector FID, 310 °C
Description
Categories Analytical Chromatography, Food & Beverage Analysis, SPE Applications, GC Applications, Phenols & Semivolatiles in water, Processing/Packaging Contaminants, PAHs, Hydrocarbons, Applications (Chromatograms)
Featured Industry Environmental
Food and Beverages
General description See "Related Products" to find associated products for sale.
For more information, email: Supelco Technical Service
Legal Information Supelclean is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE
Analyte
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[ghi]perylene
  • Benzo[k]fluoranthene
  • Chrysene
  • Dibenzo[a,h]anthracene
  • Dibenzo[a,h]anthracene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Indeno[1,2,3-cd]pyrene
  • Phenanthrene
  • Pyrene