GC Analysis of Polyunsaturated Fatty Acid (PUFA) Methyl Esters on PAG
GC Analysis of Polyunsaturated Fatty Acid (PUFA) Methyl Esters on PAG
Conditions
column PAG, 30 m x 0.25 mm I.D., 0.25 µm (24223)
oven 220 °C
carrier gas helium, 25 cm/sec @ 220 °C
sample Omegawax Test Mix (48476)
injection 1 µL, split 100:1
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • C14:0
  • C16:0
  • C18:0
  • C18:1n9
  • C18:2
  • C18:3
  • C18:4n3
  • C20:0
  • C20:1
  • C20:5n3
  • C22:0
  • C22:5n3
  • C22:6n3
  • C23:0
  • C24:0
  • C24:1
  • Methyl arachidate
  • Methyl eicosenoate
  • Methyl linoleate
  • Methyl linolenate
  • Methyl oleate
  • Methyl stearate
  • Methyl stearidonate
  • Methyl tricosanoate