GC Analysis of Potpourri Fragrance Compounds on SUPELCOWAX® 10
GC Analysis of Potpourri Fragrance Compounds on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079)
oven 50 °C (0 min), then 8 °C/min to 270 °C (35 min)
carrier gas helium, 25cm/ sec set at 110C
sample Mixture of Potpourri Fragrance Compounds, neat
injection 0.1 µL100:1 split
liner 4 mm I.D. cup injector
inj. temp. 250 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Flavors and Fragrances
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes