GC Analysis of Proline Methyl Ester Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA
GC Analysis of Proline Methyl Ester Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 155 °C
carrier gas helium, 30 psi
sample 3 mg/mL racemate in methanolpeak 1: D-proline methyl ester (N-acetyl derivative)peak 2: L-proline methyl ester (N-acetyl derivative
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Elution order reversal using the N-trifluoroacetyl derivative.
Categories Analytical Chromatography, GC Applications, Enantiomers, Amino Acids
Featured Industry Food and Beverages
Life Science and Biopharma
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes