GC Analysis of Propylene Oxide Enantiomers on Astec® CHIRALDEX® A-TA
GC Analysis of Propylene Oxide Enantiomers on Astec® CHIRALDEX® A-TA
Conditions
column Astec CHIRALDEX A-TA, 30 m x 0.25 mm I.D., 0.12 µm (73003AST)
oven 30 °C
carrier gas helium, 24 psi
sample peak 1: R(+)-propylene oxidepeak 2: S(-)-propylene oxide
injection 3 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Can also substitute CHIRALDEX B-DP for this separation.
Categories Analytical Chromatography, GC Applications, Enantiomers, Volatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes