Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on Omegawax® (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on Omegawax® (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Omegawax 100, 15 m x 0.10 mm I.D., 0.10 µm (23399-U)
oven 140 C, 40 C/min to 280 C (2 min)
carrier gas hydrogen, 50 cm/sec constant
sample PUFA No. III - Menhaden Oil (47085-U), diluted to 50 mg/mL in methylene chloride
injection 0.2 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 C
detector FID, 280 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • cis-11-Octadecenoic methyl ester
  • cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester
  • Methyl all-cis-5,8,11,14,17-eicosapentaenoate
  • Methyl all-cis-7,10,13,16,19-docosapentaenoate
  • Methyl arachidonate
  • Methyl linoleate
  • Methyl linolenate
  • Methyl linolenate
  • Methyl myristate
  • Methyl oleate
  • Methyl palmitate
  • Methyl palmitoleate
  • Methyl stearate
  • Methyl stearidonate
  • Methyl cis-11-eicosenoate
  • Methyl cis-6,9,12,15-hexadecatetraenoate
  • Methyl cis-6,9,12-hexadecatrienoate
  • Methyl cis-8,11,14,17-eicosatetraenoate
  • Methyl cis-9,12-hexadecadienoate