GC Analysis of QA Test Mix Containing FAMEs on Omegawax
GC Analysis of QA Test Mix Containing FAMEs on Omegawax
Conditions
column Omegawax 320, 30 m x 0.32 mm I.D., 0.25 µm (24152)
oven 200 °C
carrier gas helium, 25 cm/sec @ 200 °C
sample Omegawax Column Test Mix (48476), methyl ester derivatives of a naturally occurring mix of fatty acids, total concentration of 50 mg/mL in hexane
injection 1 µL, 100:1 split
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages