GC Analysis of QA Test Mix on SLB®-5ms, before Extended Hold at 360 °C
GC Analysis of QA Test Mix on SLB®-5ms, before Extended Hold at 360 °C
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 1.0 µm (28476-U)
oven 145 °C (20 min), 15 °C/min to 325 °C (15 min), 15 °C/min to 340 °C (15 min), 15 °C/min to 360 °C (4 hours)
carrier gas helium, 30 cm/sec. @ 145 °C
sample Non-polar Column Test Mix (47300-U), 500 µg/mL in methylene chloride
injection 1.0 µL100:1 split
liner 4 mm I.D. split, cup design
inj. temp. 250 °C
detector FID, 360 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1-Octanol
  • 2,6-Dimethylaniline
  • 2,6-Dimethylphenol
  • 2-Octanone
  • n-Decane
  • n-Dodecane
  • n-Tridecane
  • n-Undecane