GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column PEG, 30 m x 0.25 mm I.D., 0.25 µm
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 155 °C (25 min) for PEG 3; 130 °C (25 min) for SLB-IL60
carrier gas helium, 20 cm/sec for PEG 3; helium, 25 cm/sec for SLB-IL60
sample polar column test mix (47302), 9 analytes, each at 500 µg/mL in methylene chloride
injection 1.0 µL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1-Octanol
  • 2,6-Dimethylaniline
  • 2,6-Dimethylphenol
  • 2-Octanone
  • n-Eicosane
  • n-Heptadecane
  • n-Hexadecane
  • n-Octadecane
  • n-Pentadecane