GC Analysis of QA Test Mix on SLB®-IL60 before/after Carrier Gas Flow Stopped
GC Analysis of QA Test Mix on SLB®-IL60 before/after Carrier Gas Flow Stopped
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 130 C (25 min)
carrier gas helium, 25 cm/sec
sample polar column test mix (47302), 9 analytes, each at 500 µg/mL in methylene chloride
injection 1.0 muL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1-Octanol
  • 2,6-Dimethylaniline
  • 2,6-Dimethylphenol
  • 2-Octanone
  • n-Eicosane
  • n-Heptadecane
  • n-Hexadecane
  • n-Octadecane
  • n-Pentadecane