GC Analysis of (R)-(+)-1-Phenylethylamine (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
GC Analysis of ®-(+)-1-Phenylethylamine (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
Conditions
column Astec CHIRALDEX G-PN, 30 m x 0.25 mm I.D., 0.12 µm (74033AST)
oven 130 °C
carrier gas helium, 30 psi
sample 3 mg/mL in methanolpeak 1: ®-(+)-1-phenylethylamine (N-TFA derivative) (ee% 99.1)peak 2: (S)-(-)-1-phenylethylamine (N-TFA derivative)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Anilines, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes