GC Analysis of (R)-(+)-1-Phenylethanol on Astec® CHIRALDEX® B-PM
GC Analysis of ®-(+)-1-Phenylethanol on Astec® CHIRALDEX® B-PM
Conditions
column Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 µm (76023AST)
oven 120 °C
carrier gas helium, 24 psi
sample 3 mg/mL in methylene chloridepeak 1: ®-(+)-1-phenylethanol
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Alcohols, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • R(+)-1-Phenylethanol