Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
Conditions
column TCEP, 15 m x 0.10 mm I.D., 0.18 µm (28348-U)
oven 100 C
carrier gas hydrogen, 43 cm/sec
sample Unleaded gasoline (refinery grade)
injection 0.2 µL, 500:1 split
liner 4 mm I.D., split, cup design
inj. temp. 220 C
detector FID, 220 C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3-Trimethylbenzene
  • 1,2,3-Trimethylbenzene
  • 1,2,4-Trimethyl benzene
  • 1,2,4-Trimethylbenzene
  • Benzene
  • Benzene
  • Cumene
  • Cumene
  • Ethyl benzene
  • Ethyl toluene
  • Ethyl xylene
  • Ethylbenzene
  • Indan
  • Mesitylene
  • Mesitylene
  • Methylindan
  • m-Xylene
  • m-Xylene
  • o-Xylene
  • o-Xylene
  • Pentamethylbenzene
  • Pentamethylbenzene
  • Propyl toluene
  • Propyl toluene
  • Propylbenzene
  • Propylbenzene
  • p-Xylene
  • p-Xylene
  • Tetramethyl benzene
  • Toluene
  • Toluene
  • Tridecane
  • Tridecane