GC Analysis of Repeat Injections of a QC Test Mix Containing Water on Watercol™ 1460, 110 °C
GC Analysis of Repeat Injections of a QC Test Mix Containing Water on Watercol™ 1460, 110 °C
Conditions
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 110 C
carrier gas helium, 26 cm/sec
sample 6 analytes in acetone, each at 1 mg/mL except water (at 0.5 mg/mL)
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 C
detector TCD, 200 C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water does not interfere chromatographically with other small polar analytes. This application shows the ruggedness of Watercol 1460 to repeat injections.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC