GC Analysis of Scotch Whiskey on a Packed Column (5% Carbowax® 20M on 80/120 Carbopack B)
GC Analysis of Scotch Whiskey on a Packed Column (5% Carbowax® 20M on 80/120 °Carbopack B)
Conditions
column 2 m x 2 mm I.D. glass
packing 5% Carbowax 20M on 80/120 °Carbopack B
oven 70 °C, 4C/min. to 150 °C
carrier gas nitrogen, 20 mL/min
sample Scotch Whiskey
injection 1 µL
detector FID
Description
Categories Analytical Chromatography, Food & Beverage Analysis, GC Applications, Beverage Testing, Beer, Wine, and Spirits, Alcohols, Spirits
Featured Industry Food and Beverages
Legal Information Carbowax is a registered trademark of Union Carbide
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Propanol
  • Acetaldehyde
  • Acetic acid
  • Active amyl alcohol
  • Ethanol
  • Ethyl acetate
  • Isoamyl alcohol
  • Isobutanol
  • Methanol