GC Analysis of Short and Long Chain Free Fatty Acids on Nukol™
GC Analysis of Short and Long Chain Free Fatty Acids on Nukol™
Conditions
column Nukol, 15 m x 0.53 mm I.D., 0.50 µm (25326)
oven 100 C, 10 C/min to 220 C
carrier gas helium, 30 ml/min
sample 16 analytes, at various concentrations from 50 to 800 µg/mL
injection 0.5 µL, direct injection
detector FID
Description
Categories Analytical Chromatography, GC Applications, Acids, Fatty Acids, Organic Acids
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Nukol is a trademark of Sigma-Aldrich Co. LLC